trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3,4,5-Trifluoro-4'-((trans,trans)-4'-propyl-[1,1'-bi(cyclohexan)]-4-yl)-1,1'-biphenyl
T3319-1G
T3319-5G
TCT3319-1GEA
54.4
USD
TCT3319-1G
TCT3319-5G
trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
trans,trans-3,4,5-Trifluoro-4'-(4'-propylbicyclohexyl-4-yl)biphenyl
Formula:
C₂₇H₃₃F₃ MW: 414.56 g/mol |
MDL Number:
MFCD13188632 CAS Number: 137529-41-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Mesomorphic range | 101.0 to 260.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...