2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,3-Difluoro-1-methoxy-4-[(trans-4-propylcyclohexyl)methoxy]benzene
D4909-1G
D4909-5G
TCD4909-1GEA
77
USD
TCD4909-1G
TCD4909-5G
2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole ≥98.0% (by GC)
2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole
Formula:
C₁₇H₂₄F₂O₂ MW: 298.37 g/mol |
CAS Number:
1373116-00-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 50.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...