Trimethylsilyl methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

T3293-25ML T3293-5ML
TCT3293-25MLEA 146 USD
TCT3293-25ML TCT3293-5ML
Trimethylsilyl methacrylate ≥98.0% (by GC) stabilized
Trimethylsilyl methacrylate
Formula: C₇H₁₄O₂Si
Boiling Pt: 52°C
Flash Pt: 32°C
MDL Number: MFCD00053868
CAS Number: 13688-56-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR