3,5-Dichloro-4-pyridinecarboxaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5790-1G
D5790-5G
TCD5790-1GEA
27
USD
TCD5790-1G
TCD5790-5G
3,5-Dichloro-4-pyridinecarboxaldehyde ≥98.0% (by GC)
3,5-Dichloro-4-pyridinecarboxaldehyde
Formula:
C₆H₃Cl₂NO |
MDL Number:
MFCD04039921 CAS Number: 136590-83-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...