4,4'-Dihexyl-2,2'-bithiophene ≥95.0% (by GC)

Supplier: TCI America

D4182-1G D4182-200MG
TCD4182-1GEA 445.7 USD
TCD4182-1G TCD4182-200MG
4,4'-Dihexyl-2,2'-bithiophene ≥95.0% (by GC)
4,4'-Dihexyl-2,2'-bithiophene
Formula: C₂₀H₃₀S₂
Storage Temperature: Refrigerator
CAS Number: 135926-94-2

Order Now

Specification Test Results

Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR