2-Acetamidobenzaldehyde 98,0 GC_ASSAY_METHOD

Supplier: TCI America

A2926-1G A2926-200MG
TCA2926-1GEA 507 USD
TCA2926-1G TCA2926-200MG
2-Acetamidobenzaldehyde 98,0 GC_ASSAY_METHOD
2-Acetamidobenzaldehyde
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 103°C
MDL Number: MFCD06739581
CAS Number: 13493-47-5

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR