4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole ≥98.0% (by GC)

Supplier: TCI America

D4750-1G D4750-200MG
TCD4750-1GEA 183 USD
TCD4750-1G TCD4750-200MG
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole ≥98.0% (by GC)
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole
Formula: C₆HBr₂FN₂S
MW: 311.95 g/mol
MDL Number: MFCD26939241
CAS Number: 1347736-74-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR