2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

P2562-1G P2562-200MG
76220-458EA 260 USD
76220-458 76220-460
2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₃₁H₂₇BO₂
MDL Number: MFCD30187274
CAS Number: 1346007-05-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 214.0 to 218.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR