3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4979-1G
D4979-200MG
TCD4979-1GEA
257
USD
TCD4979-1G
TCD4979-200MG
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole
Formula:
C₂₄H₁₅Br₂N MW: 477.20 g/mol |
MDL Number:
MFCD22571691 CAS Number: 1345021-52-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...