9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D4919-1G
D4919-200MG
TCD4919-1GEA
422
USD
TCD4919-1G
TCD4919-200MG
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole
Formula:
C₃₀H₁₉NO |
MDL Number:
MFCD30478530 CAS Number: 1338446-77-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...