Lercanidipine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Lercanidipine HCl, 3-(1-((3,3-Diphenylpropyl)(methyl)amino)-2-methylpropan-2-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride

L0282-1G L0282-200MG
TCL0282-1GEA 45 USD
TCL0282-1G TCL0282-200MG
Lercanidipine hydrochloride ≥98.0% (by HPLC, titration analysis)
Lercanidipine hydrochloride
Formula: C₃₆H₄₁N₃O₆·HCl
MW: 648.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07773089
CAS Number: 132866-11-6
Merck Index: 14,05446

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR