Blonanserin ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine

B4565-100MG B4565-25MG
TCB4565-100MGEA 85.8 USD
TCB4565-100MG TCB4565-25MG
Blonanserin ≥98.0% (by HPLC)
Blonanserin
Formula: C₂₃H₃₀FN₃
MW: 367.51 g/mol
MDL Number: MFCD00893838
CAS Number: 132810-10-7
Merck Index: 14,01319

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 10.80 to 12.00 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR