Dicyclohexylamine Borane ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Borane - Dicyclohexylamine Complex

B5545-25G B5545-5G
TCB5545-25GEA 163 USD
TCB5545-25G TCB5545-5G
Dicyclohexylamine Borane ≥95.0% (by titrimetric analysis)
Dicyclohexylamine Borane
Formula: BH₃·C₁₂H₂₃N
MW: 195.16 g/mol
Storage Temperature: Refrigerator
CAS Number: 131765-96-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR