2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D5270-1G D5270-200MG
76219-270EA 185 USD
76219-270 76219-272
2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole ≥98.0% (by HPLC)
2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole
Formula: C₁₈H₁₀Br₃N
MW: 480.00 g/mol
Melting Pt: 136°C
MDL Number: MFCD30469057
CAS Number: 1313900-20-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR