Ethyl 3-(3,4-Dichlorophenyl)carbazate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(3,4-Dichlorophenyl)-2-(ethoxycarbonyl)hydrazine, 3-(3,4-Dichlorophenyl)carbazic acid Ethyl ester

E0998-1G E0998-5G
TCE0998-1GEA 80 USD
TCE0998-1G TCE0998-5G
Ethyl 3-(3,4-Dichlorophenyl)carbazate ≥98.0% (by GC)
Ethyl 3-(3,4-Dichlorophenyl)carbazate
Formula: C₉H₁₀Cl₂N₂O₂
MW: 249.09 g/mol
MDL Number: MFCD28975114
CAS Number: 13124-15-7

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR