N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine tetrahydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B4478-1G
B4478-5G
TCB4478-1GEA
74
USD
TCB4478-1G
TCB4478-5G
N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine tetrahydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
N,N'-Bis(2-pyridylmethyl)-1,2-ethylenediamine tetrahydrochloride dihydrate
Formula:
C₁₄H₁₈N₄·₄HCl·2H₂O |
CAS Number:
1310544-60-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 219.0 to 223.0 deg-C |
Water | 7.0 to 10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...