Ethyl (2-Methoxyphenoxy)acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (2-Methoxyphenoxy)acetic acid Ethyl ester, Ethyl 2-(2-methoxyphenoxy)acetate

E1241-25G E1241-5G
TCE1241-25GEA 345 USD
TCE1241-25G TCE1241-5G
Ethyl (2-Methoxyphenoxy)acetate ≥98.0% (by GC)
Ethyl (2-Methoxyphenoxy)acetate
Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 116°C
MDL Number: MFCD00082657
CAS Number: 13078-21-2
EINECS: 235-977-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR