3-Benzyloxy-5-bromopyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B4775-1G
B4775-5G
TCB4775-1GEA
74.3
USD
TCB4775-1G
TCB4775-5G
3-Benzyloxy-5-bromopyridine ≥98.0% (by GC, titration analysis)
3-Benzyloxy-5-bromopyridine
Formula:
C₁₂H₁₀BrNO |
MDL Number:
MFCD07375004 CAS Number: 130722-95-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...