4,4'-(Ethyne-1,2-diyl)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5,5'-(Ethyne-1,2-diyl)bis(isobenzofuran-1,3-dione)
E1164-25G
E1164-5G
TCE1164-25GEA
222.85
USD
TCE1164-25G
TCE1164-5G
4,4'-(Ethyne-1,2-diyl)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)
4,4'-(Ethyne-1,2-diyl)diphthalic anhydride
Formula:
C₁₈H₆O₆ MW: 318.24 g/mol |
MDL Number:
MFCD28976167 CAS Number: 129808-00-0 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Morpholine Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...