trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(trans,trans)-4-(But-3-en-1-yl)-4'-(p-tolyl)-1,1'-bi(cyclohexane), trans,trans-4'-(3-Butenyl)-4-(4-methylphenyl)bicyclohexyl
B4915-1G
B4915-5G
TCB4915-1GEA
34.5
USD
TCB4915-1G
TCB4915-5G
trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)
trans,trans-4'-(3-Butenyl)-4-(p-tolyl)bicyclohexyl
Formula:
C₂₃H₃₄ |
MDL Number:
MFCD13188633 CAS Number: 129738-42-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...