5-Azoniaspiro[4.4]nonane Hexafluorophosphate ≥96.0% (by total nitrogen basis)

Supplier: TCI America
Warning

A3102-1G A3102-5G
TCA3102-1GEA 82 USD
TCA3102-1G TCA3102-5G
5-Azoniaspiro[4.4]nonane Hexafluorophosphate ≥96.0% (by total nitrogen basis)
5-Azoniaspiro[4.4]nonane Hexafluorophosphate
Formula: C₈H₁₆F₆NP
MW: 271.19 g/mol
CAS Number: 129211-50-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR