Sodium bis(nonafluorobutanesulfonyl)imide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Bis(nonafluorobutanesulphonyl)imide sodium salt

O0492-1G O0492-5G
TCO0492-1GEA 91 USD
TCO0492-1G TCO0492-5G
Sodium bis(nonafluorobutanesulfonyl)imide ≥97.0% (by titrimetric analysis)
Sodium bis(nonafluorobutanesulfonyl)imide
Formula: C₈F₁₈NNaO₄S₂
CAS Number: 129135-86-0
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR