Ospemifene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (Z)-2-(4-(4-Chloro-1,2-diphenylbut-1-en-1-yl)phenoxy)ethan-1-ol

O0441-100MG O0441-1G
TCO0441-100MGEA 55 USD
TCO0441-100MG TCO0441-1G
Ospemifene ≥98.0% (by GC)
Ospemifene
Formula: C₂₄H₂₃ClO₂
MW: 378.90 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00871890
CAS Number: 128607-22-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR