2-Amino-9,9-diphenylfluorene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
A2779-1G
A2779-5G
TCA2779-1GEA
94.2
USD
TCA2779-1G
TCA2779-5G
2-Amino-9,9-diphenylfluorene ≥98.0% (by HPLC, titration analysis)
2-Amino-9,9-diphenylfluorene
Formula:
C₂₅H₁₉N |
MDL Number:
MFCD22376664 CAS Number: 1268519-74-9 |
Specification Test Results
Appearance | White to Yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...