4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(2-Methyl-1-propenyl)boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(2-methylprop-1-en-1-yl)-1,3,2-dioxaborolane
T3469-1G
T3469-5G
TCT3469-1GEA
60
USD
TCT3469-1G
TCT3469-5G
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane ≥98.0% (by GC)
4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane
Formula:
C₁₀H₁₉BO₂ MW: 182.07 g/mol Boiling Pt: 71°C Flash Pt: 66°C |
MDL Number:
MFCD09842762 CAS Number: 126689-00-7 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Specific gravity (20/20) | 0.8960 to 0.9000 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...