9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9-Phenyl-3,6-bis(4-(1-phenyl-1H-benzo[d]imidazol-2-yl)phenyl)-9H-carbazole

P2330-1G P2330-200MG
TCP2330-1GEA 443 USD
TCP2330-1G TCP2330-200MG
9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole ≥98.0% (by HPLC)
9-Phenyl-3,6-bis[4-(1-phenylbenzimidazol-2-yl)phenyl]carbazole
Formula: C₅₆H₃₇N₅
Melting Pt: 209°C
MDL Number: MFCD31618148
CAS Number: 1258780-50-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR