4-(1-Propynyl)phthalic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(Methylethynyl)phthalic anhydride

M2757-25G M2757-5G
TCM2757-25GEA 145.48 USD
TCM2757-25G TCM2757-5G
4-(1-Propynyl)phthalic anhydride ≥98.0% (by GC, titration analysis)
4-(1-Propynyl)phthalic anhydride
Formula: C₁₁H₆O₃
MW: 186.17 g/mol
CAS Number: 1240685-26-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR