1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
M2998-1G
M2998-5G
76220-322EA
111
USD
76220-322
76220-324
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulfonyl)imide
Formula:
C₈H₁₈F₂N₂O₅S₂ MW: 324.36 g/mol |
MDL Number:
MFCD30749293 CAS Number: 1235234-47-5 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...