3-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

B4888-1G B4888-5G
TCB4888-1GEA 38.7 USD
TCB4888-1G TCB4888-5G
3-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
3-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formula: C₂₄H₁₇Br
MDL Number: MFCD28134530
CAS Number: 1233200-57-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR