Itopride hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
I0948-5G
I0948-1G
TCI0948-5GEA
83
USD
TCI0948-5G
TCI0948-1G
Itopride hydrochloride ≥98.0% (by HPLC, titration analysis)
Itopride hydrochloride
Formula:
C₂₀H₂₆N₂O₄·HCl Storage Temperature: Refrigerator |
MDL Number:
MFCD00881710 CAS Number: 122892-31-3 Merck Index: 14,05244 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 192.0 to 196.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...