N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: MeO-TPD

T3290-1G T3290-200MG
TCT3290-1GEA 235 USD
TCT3290-1G TCT3290-200MG
N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetrakis(4-methoxyphenyl)benzidine
Formula: C₄₀H₃₆N₂O₄
MW: 608.74 g/mol
MDL Number: MFCD09833415
CAS Number: 122738-21-0

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR