3-Bromo-9-(4-bromophenyl)-9H-carbazole ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
B5107-1G
B5107-5G
TCB5107-1GEA
154
USD
TCB5107-1G
TCB5107-5G
3-Bromo-9-(4-bromophenyl)-9H-carbazole ≥97.0% (by GC)
3-Bromo-9-(4-bromophenyl)-9H-carbazole
Formula:
C₁₈H₁₁Br₂N MW: 401.10 g/mol |
MDL Number:
MFCD30063199 CAS Number: 1226860-66-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...