10,10'-Dibromo-9,9'-bianthracene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4254-1G D4254-5G
TCD4254-1GEA 138.1 USD
TCD4254-1G TCD4254-5G
10,10'-Dibromo-9,9'-bianthracene ≥98.0% (by HPLC, titration analysis)
10,10'-Dibromo-9,9'-bianthracene
Formula: C₂₈H₁₆Br₂
MW: 512.24 g/mol
Melting Pt: 342°C
MDL Number: MFCD11045035
CAS Number: 121848-75-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR