(1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
GO-Y078
H1525-100MG
H1525-20MG
76220-128EA
334
USD
76220-128
76220-130
(1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one ≥95.0% (by HPLC)
(1E,4E)-1-(4-Hydroxy-3,5-dimethoxyphenyl)-5-(3,4,5-trimethoxyphenyl)-1,4-pentadiene-3-one
Formula:
C₂₂H₂₄O₇ MW: 400.43 g/mol |
CAS Number:
1217503-60-0 EINECS: 274-722-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...