3-Cyclohexylthiophene ≥96.0% (by GC)

Supplier: TCI America

C3400-1G C3400-5G
TCC3400-1GEA 173 USD
TCC3400-1G TCC3400-5G
3-Cyclohexylthiophene ≥96.0% (by GC)
3-Cyclohexylthiophene
Formula: C₁₀H₁₄S
MW: 166.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02260242
CAS Number: 120659-34-9

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR