2-Bromo-8-iododibenzothiophene ≥97.0% (by GC)

Supplier: TCI America

B4904-1G B4904-200MG
TCB4904-1GEA 500.1 USD
TCB4904-1G TCB4904-200MG
2-Bromo-8-iododibenzothiophene ≥97.0% (by GC)
2-Bromo-8-iododibenzothiophene
Formula: C₁₂H₆BrIS
MW: 389.05 g/mol
MDL Number: MFCD28975102
CAS Number: 1206544-88-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 209.0 to 213.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR