(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2909-1G M2909-5G
76220-300EA 120 USD
76220-300 76220-302
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
(4-Methylphenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate
Formula: C₁₇H₁₈F₃IO₃S
MDL Number: MFCD20264882
CAS Number: 1204518-02-4

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 177.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR