2-Amino-4'-chlorobiphenyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4'-Chloro-[1,1'-biphenyl]-2-amine

A3034-1G A3034-5G
TCA3034-1GEA 134 USD
TCA3034-1G TCA3034-5G
2-Amino-4'-chlorobiphenyl ≥98.0% (by GC, titration analysis)
2-Amino-4'-chlorobiphenyl
Formula: C₁₂H₁₀ClN
MW: 203.67 g/mol
MDL Number: MFCD03424659
CAS Number: 1204-44-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR