9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4952-1G B4952-200MG
TCB4952-1GEA 256 USD
TCB4952-1G TCB4952-200MG
9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene ≥98.0% (by GC)
9-Bromo-7,7-dimethyl-7H-benzo[c]fluorene
Formula: C₁₉H₁₅Br
MW: 323.23 g/mol
MDL Number: MFCD27980277
CAS Number: 1198396-46-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR