3-(2-Bromophenyl)-9-phenyl-9H-carbazole ≥97.0% (by GC)

Supplier: TCI America
Warning

B5465-1G B5465-5G
TCB5465-1GEA 174 USD
TCB5465-1G TCB5465-5G
3-(2-Bromophenyl)-9-phenyl-9H-carbazole ≥97.0% (by GC)
3-(2-Bromophenyl)-9-phenyl-9H-carbazole
Formula: C₂₄H₁₆BrN
MW: 398.30 g/mol
MDL Number: MFCD29071836
CAS Number: 1190100-35-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR