2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene ≥99.0% (by HPLC)

Supplier: TCI America

D4862-1G D4862-200MG
76219-728EA 447 USD
76219-728 76219-730
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene ≥99.0% (by HPLC)
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene
Formula: C₃₄H₃₂Br₂
CAS Number: 1187763-68-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR