2-(4-Bromophenyl)-5-phenylthiophene ≥98.0% (by GC)

Supplier: TCI America

B5491-1G B5491-200MG
76219-634EA 214 USD
76219-634 76219-636
2-(4-Bromophenyl)-5-phenylthiophene ≥98.0% (by GC)
2-(4-Bromophenyl)-5-phenylthiophene
Formula: C₁₆H₁₁BrS
MW: 315.23 g/mol
MDL Number: MFCD21648452
CAS Number: 118621-30-0

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR