1,2-Difluoro-4-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (by GC)

Supplier: TCI America

D5128-1G D5128-5G
76219-168EA 121 USD
76219-168 76219-170
1,2-Difluoro-4-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (by GC)
1,2-Difluoro-4-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene
Formula: C₂₃H₃₄F₂
MW: 348.52 g/mol
MDL Number: MFCD12912373
CAS Number: 117943-37-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 37 to 107 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR