7-(Diethylamino)-3-(2-thienyl)coumarin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

D5007-1G D5007-5G
TCD5007-1GEA 157 USD
TCD5007-1G TCD5007-5G
7-(Diethylamino)-3-(2-thienyl)coumarin ≥98.0% (by HPLC, titration analysis)
7-(Diethylamino)-3-(2-thienyl)coumarin
Formula: C₁₇H₁₇NO₂S
MW: 299.39 g/mol
CAS Number: 117850-52-9

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Lambda max. 420.0 to 425.0 nm (MeOH)
Absorbance (E1%1cm) min. 1150 (MeOH, 420.0 to 425.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR