1-(Trifluoromethanesulfonyl)-1H-benzotriazole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3167-1G T3167-5G
76220-536EA 142 USD
76220-536 76220-538
1-(Trifluoromethanesulfonyl)-1H-benzotriazole ≥98.0% (by HPLC)
1-(Trifluoromethanesulfonyl)-1H-benzotriazole
Formula: C₇H₄F₃N₃O₂S
MW: 251.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00963686
CAS Number: 117632-84-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 36.0 to 40.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR