1-Methylpyridinium Hexafluorophosphate ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
M3195-25G
M3195-5G
TCM3195-25GEA
269
USD
TCM3195-25G
TCM3195-5G
1-Methylpyridinium Hexafluorophosphate ≥98.0% (by HPLC)
1-Methylpyridinium Hexafluorophosphate
Formula:
C₆H₈F₆NP |
CAS Number:
117029-34-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...