4-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-Bromo-5'-phenyl-m-terphenyl, 5'-(4-Bromophenyl)-1,1':3',1''-terphenyl, 5'-(4-Bromophenyl)-m-terphenyl
B4889-1G
B4889-5G
TCB4889-1GEA
487
USD
TCB4889-1G
TCB4889-5G
4-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
4-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formula:
C₂₄H₁₇Br MW: 385.30 g/mol |
MDL Number:
MFCD26407506 CAS Number: 116941-52-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...