4-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)
Supplier: TCI America
B5031-1G
B5031-5G
TCB5031-1GEA
37
USD
TCB5031-1G
TCB5031-5G
4-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)
4-Bromo-9,9'-spirobi[9H-fluorene]
Formula:
C₂₅H₁₅Br |
MDL Number:
MFCD16660920 CAS Number: 1161009-88-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...