9-(4-Bromophenyl)-10-(1-naphthyl)anthracene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B5029-1G
B5029-200MG
TCB5029-1GEA
331
USD
TCB5029-1G
TCB5029-200MG
9-(4-Bromophenyl)-10-(1-naphthyl)anthracene ≥98.0% (by HPLC, titration analysis)
9-(4-Bromophenyl)-10-(1-naphthyl)anthracene
Formula:
C₃₀H₁₉Br MW: 459.39 g/mol |
MDL Number:
MFCD26400490 CAS Number: 1160506-32-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...