2-(4-Bromophenyl)triphenylene ≥98.0% (by GC)
Supplier: TCI America
B4955-1G
B4955-5G
TCB4955-1GEA
212.4
USD
TCB4955-1G
TCB4955-5G
2-(4-Bromophenyl)triphenylene ≥98.0% (by GC)
2-(4-Bromophenyl)triphenylene
Formula:
C₂₄H₁₅Br |
CAS Number:
1158227-56-5 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...